(R)-(-)-1-Aminopropan-2-ol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
CAS Number: 2799-16-8
MDL Number: MFCD00064428
Molecular Formula: C3H9NO
Molecular Weight: 75.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 160
Flash Point (°C): 78
Freezing point (°C): 24
Specific rotation [a]20/D: -24 deg (C=1, MeOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-Isopropanolamine
Formula:
C₃H₉NO Boiling Pt: 160 °C Flash Pt: 78 °C |
MDL Number:
MFCD00064428 CAS Number: 2799-16-8 UN: 3259 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...