(S)-(+)-2-Amino-1-butanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: D(+)-2-Amino-1-butanol, (S)-2-Aminobutan-1-ol

A0973-1ML A0973-5ML
TCA0973-1MLEA 34 USD
TCA0973-1ML TCA0973-5ML
(S)-(+)-2-Amino-1-butanol ≥98.0% (by GC, titration analysis)
(S)-(+)-2-Amino-1-butanol

CAS Number: 5856-62-2
MDL Number: MFCD00064418
Molecular Formula: C4H11NO
Molecular Weight: 89.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 178
Flash Point (°C): 79
Specific Gravity (20/20): 0.94
Specific rotation [a]20/D: 9.5 deg (neat)

Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 178 °C
Density: 0.94 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00064418
CAS Number: 5856-62-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec8d9dea66693f185fd2eb51717b782058f8793b&vid=ec8d9dea66693f185fd2eb51717b782058f8793b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR