4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole ≥95.0% (by titrimetric analysis)
Supplier: TCI America
A0956-25G
A0956-5G
TCA0956-25GEA
248
USD
TCA0956-25G
TCA0956-5G
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole ≥95.0% (by titrimetric analysis)
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole
[for Determination of Aldehydes]
CAS Number: 1750-12-5
MDL Number: MFCD00003098
Molecular Formula: C2H6N6S
Molecular Weight: 146.17
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Melting point (°C): 240
Formula:
C₂H₆N₆S Melting Pt: 240 °C |
MDL Number:
MFCD00003098 CAS Number: 1750-12-5 UN: 1325 ADR: 4.1,III |
|
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...