(±)-2-Amino-1-propanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: DL-Alaninol, 2-aminopropan-1-ol, DL-2-Amino-1-propanol

A0942-25G A0942-5G
TCA0942-25GEA 81 USD
TCA0942-25G TCA0942-5G
(±)-2-Amino-1-propanol ≥98.0% (by GC, titration analysis)
(±)-2-Amino-1-propanol

CAS Number: 6168-72-5
MDL Number: MFCD00008085
Molecular Formula: C3H9NO
Molecular Weight: 75.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 175
Flash Point (°C): 83
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: DL-Alaninol

Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 175 °C
Density: 0.96 (20 °C)
Flash Pt: 83 °C
MDL Number: MFCD00008085
CAS Number: 6168-72-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81fc014e12b40b8199bfeff6364f386246918d73&vid=81fc014e12b40b8199bfeff6364f386246918d73&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR