3-Amino-2,2-dimethyl-1-propanol ≥97.0% (by GC, titration analysis)
Supplier: TCI America
A0914-25G
A0914-5G
TCA0914-25GEA
277.2
USD
TCA0914-25G
TCA0914-5G
3-Amino-2,2-dimethyl-1-propanol ≥97.0% (by GC, titration analysis)
3-Amino-2,2-dimethyl-1-propanol
CAS Number: 26734-09-8
MDL Number: MFCD00059847
Molecular Formula: C5H13NO
Molecular Weight: 103.17
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Other Product Names / Synonyms: Neopentanolamine
Formula:
C₅H₁₃NO MW: 103.17 g/mol |
MDL Number:
MFCD00059847 CAS Number: 26734-09-8 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...