1-(2-Aminoethyl)pyrrolidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
A0884-25G
A0884-5G
TCA0884-25GEA
246
USD
TCA0884-25G
TCA0884-5G
1-(2-Aminoethyl)pyrrolidine ≥98.0% (by GC, titration analysis)
CAS Number: 7154-73-6
MDL Number: MFCD00003182
Molecular Formula: C6H14N2
Molecular Weight: 114.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 161
Flash Point (°C): 47
Specific Gravity (20/20): 0.91
Formula:
C₆H₁₄N₂ Boiling Pt: 161 °C Density: 0.91 (20 °C) Flash Pt: 47 °C |
MDL Number:
MFCD00003182 CAS Number: 7154-73-6 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...