5-Aminopentan-1-ol ≥95.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
5-Amino-1-pentanol
A0875-250G
A0875-25G
TCA0875-250GEA
792
USD
TCA0875-250G
TCA0875-25G
5-Aminopentan-1-ol ≥95.0% (by GC, titration analysis)
5-Aminopentan-1-ol
CAS Number: 2508-29-4
MDL Number: MFCD00008237
Molecular Formula: C5H13NO
Molecular Weight: 103.17
Purity/Analysis Method: >95.0% (GC,T)
Form: Crystal
Boiling point (°C): 222
Flash Point (°C): 65
Freezing point (°C): 36
Formula:
C₅H₁₃NO MW: 103.16 g/mol Boiling Pt: 222 °C Flash Pt: 65 °C |
MDL Number:
MFCD00008237 CAS Number: 2508-29-4 UN: 3259 ADR: 8,III |
Specification Test Results
Purity/Analysis Method | >95.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...