4-Aminomorpholine ≥98.0%

Supplier: TCI America

Synonyms: N-Aminomorpholine

A0832-25ML A0832-5ML
TCA0832-25MLEA 97 USD
TCA0832-25ML TCA0832-5ML
4-Aminomorpholine ≥98.0%
4-Aminomorpholine

CAS Number: 4319-49-7
MDL Number: MFCD00006174
Molecular Formula: C4H10N2O
Molecular Weight: 102.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 168
Flash Point (°C): 58
Specific Gravity (20/20): 1.06

Formula: C₄H₁₀N₂O
Boiling Pt: 168 °C
Density: 1.06 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00006174
CAS Number: 4319-49-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bcc609e6b14b5ce935d6ae5002ee9c6595392ee0&vid=bcc609e6b14b5ce935d6ae5002ee9c6595392ee0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR