5-Amino-1H-indazole ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 1H-Indazol-5-amine, Indazol-5-amine, 5-Amino-1H-indazole, 5-Aminoindazole, 1H-Indazol-5-ylamine, Indazol-5-ylamine

A0797-25G A0797-5G
TCA0797-25GEA 247.9 USD
TCA0797-25G TCA0797-5G
5-Amino-1H-indazole ≥98.0% (by HPLC, titration analysis)
5-Amino-1H-indazole

CAS Number: 19335-11-6
MDL Number: MFCD00037975
Molecular Formula: C7H7N3
Molecular Weight: 133.15
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 177

Formula: C₇H₇N₃
MW: 133.15 g/mol
Melting Pt: 177 °C
MDL Number: MFCD00037975
CAS Number: 19335-11-6
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a35335b28ae078ca851ab1c6f8106a2751d2123&vid=8a35335b28ae078ca851ab1c6f8106a2751d2123&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR