(±)-3,3-Dimethyl-2-butylamine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1,2,2-Trimethylpropylamine, 3,3-Dimethylbutan-2-amine

A0656-10ML A0656-25ML
TCA0656-10MLEA 238.5 USD
TCA0656-10ML TCA0656-25ML
(±)-3,3-Dimethyl-2-butylamine ≥98.0% (by GC)
(±)-3,3-Dimethyl-2-butylamine

CAS Number: 3850-30-4
MDL Number: MFCD00008078
Molecular Formula: C6H15N
Molecular Weight: 101.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 102
Flash Point (°C): 1
Specific Gravity (20/20): 0.77


Other Product Names / Synonyms: 3-Amino-2,2-dimethylbutane, 2-Amino-3,3-dimethylbutane, 1,2,2-Trimethylpropylamine

Formula: C₆H₁₅N
MW: 101.19 g/mol
Boiling Pt: 102 °C
Density: 0.77 (20 °C)
Flash Pt: 1 °C
MDL Number: MFCD00008078
CAS Number: 3850-30-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f2ea619d7a3287fc6ee0dc77b3e3ed83a76d81c&vid=3f2ea619d7a3287fc6ee0dc77b3e3ed83a76d81c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR