Acrolein diethyl acetal ≥97.0%

Supplier: TCI America

Synonyms: Acrolein Acetal, 1,1-Diethoxy-2-propene, 3,3-Diethoxy-1-propene

A0640-250G A0640-25G
TCA0640-250GEA 467 USD
TCA0640-250G TCA0640-25G
Acrolein diethyl acetal ≥97.0%
Acrolein diethyl acetal

CAS Number: 3054-95-3
MDL Number: MFCD00009239
Molecular Formula: C7H14O2
Molecular Weight: 130.19
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 124
Flash Point (°C): 15
Specific Gravity (20/20): 0.85


Other Product Names / Synonyms: Acrolein Acetal, 1,1-Diethoxy-2-propene, 3,3-Diethoxy-1-propene

Formula: C₇H₁₄O₂
Boiling Pt: 124 °C
Density: 0.85 (20 °C)
Flash Pt: 15 °C
MDL Number: MFCD00009239
CAS Number: 3054-95-3
UN: 2374
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d01503324f781a977ef0cc7638c4c4560667c749&vid=7176f6fba7ca02b12dc629e243c2e9c0a6deb599&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR