Aluminium triethanolate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Aluminumethoxide, Aluminum ethoxide, Aluminium triethylate

A0245-250G A0245-25G
TCA0245-250GEA 149 USD
TCA0245-250G TCA0245-25G
Aluminium triethanolate ≥98.0% (by titrimetric analysis)
Aluminium triethanolate

CAS Number: 555-75-9
MDL Number: MFCD00009658
Molecular Formula: C6H15AlO3
Molecular Weight: 162.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 200
Melting point (°C): 140
Flash Point (°C): 30

Formula: C₆H₁₅AlO₃
MW: 162.16 g/mol
Boiling Pt: 200 °C
Melting Pt: 140 °C
Flash Pt: 30 °C
MDL Number: MFCD00009658
CAS Number: 555-75-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26e3da3d19032ee96c2a013089d94bbbeecf9e81&vid=26e3da3d19032ee96c2a013089d94bbbeecf9e81&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR