Acetylthiocholine iodide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (2-Mercaptoethyl)trimethylammonium iodide acetate, 2-(Acetylthio)-N,N,N-trimethylethan-1-aminium iodide, S-Acetylthiocholine iodide

A0116-1G A0116-25G A0116-5G
TCA0116-1GEA 32.56 USD
TCA0116-1G TCA0116-25G TCA0116-5G
Acetylthiocholine iodide ≥98.0% (by titrimetric analysis)
Acetylthiocholine iodide

CAS Number: 1866-15-5
MDL Number: MFCD00011819
Molecular Formula: C7H16INOS
Molecular Weight: 289.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 206

Formula: H₃CC(S)O(CH₂)₂N(CH₃)₃I
MW: 289.18 g/mol
Melting Pt: 206 °C
MDL Number: MFCD00011819
CAS Number: 1866-15-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87d3f4d84d5d3b8bbe65be5d76730894b6566ecc&vid=87d3f4d84d5d3b8bbe65be5d76730894b6566ecc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR