Anti-Mouse Monoclonal Antibody (CF™ 488A)

Supplier: Biotium

20210 20210-1
97065-660EA 279.11 USD
97065-660 71004-198
Anti-Mouse Monoclonal Antibody (CF™ 488A)
Antibodies
Type: Secondary
Antigen: Fluorescein
Clonality: Monoclonal
Clone:
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: [delete]
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR