Anti-IgG Chicken Polyclonal Antibody (CF™ 543)

Supplier: Biotium

20334-500UL 20334-50UL
89427-166EA 217.08 USD
89427-166 89427-168
Anti-IgG Chicken Polyclonal Antibody (CF™ 543)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 543
Epitope: Heavy and Light chain
Host: Chicken
Isotype: IgY
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR