Anti-IgG2a Goat Polyclonal Antibody (CF™ 543)

Supplier: Biotium

20326 20326-1
89411-084EA 279.11 USD
89411-084 89411-532
Anti-IgG2a Goat Polyclonal Antibody (CF™ 543)
Antibodies
Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: CF™ 543
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR