Anti-IgG Goat Polyclonal Antibody (CF™ 543)

Supplier: Biotium

20324 20324-1
89411-080EA 217.08 USD
89411-080 89411-528
Anti-IgG Goat Polyclonal Antibody (CF™ 543)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 543
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR