Anti-IgG Donkey Polyclonal Antibody (CF™ 543)

Supplier: Biotium

20318 20318-1
89411-068EA 260.49 USD
89411-068 89411-516
Anti-IgG Donkey Polyclonal Antibody (CF™ 543)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 543
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR