Anti-IgY Donkey Polyclonal Antibody (CF™ 543)

Supplier: Biotium

20310 20310-1
89411-052EA 286.57 USD
89411-052 89411-500
Anti-IgY Donkey Polyclonal Antibody (CF™ 543)
Antibodies
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: CF™ 543
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR