Anti-TNF-a Armenian Hamster Monoclonal Antibody [clone: TN3-19.12]

Supplier: Biolegend
506101 506102
506101-BLEA 113.7 USD
506101-BL 506102-BL
Anti-TNF-a Armenian Hamster Monoclonal Antibody [clone: TN3-19.12]
Antibodies
Purified anti-mouse/rat TNF-α [TN3-19.12]; Isotype: Armenian Hamster IgG; Reactivity: Mouse, Rat, Cross-Reactivity: Rabbit (Lapine); Apps: ELISA Capture, IP, WB; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA capture applications, a concentration range of 2-6 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of mouse TNF-α recombinant protein ranging from 500 to 4 pg/ml are recommended for each ELISA plate or serial dilutions of rat TNF-α recombinant protein ranging from 2500 to 20 pg/ml are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TNF-a
Clonality: Monoclonal
Clone: TN3-19.12
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse;Rat;Cross-Reactivity: Rabbit (Lapine)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR