Anti-Histone H3 Phospho (Ser10) Mouse Monoclonal Antibody [clone: 11D8]

Supplier: Biolegend
650801 650802
650801-BLEA 198.1 USD
650801-BL 650802-BL
Anti-Histone H3 Phospho (Ser10) Mouse Monoclonal Antibody [clone: 11D8]
Antibodies
Purified anti-Histone H3 Phospho (Ser10) [11D8]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: WB; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 1 μg per ml antibody dilution buffer for each mini-gel. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Histone H3 Phospho (Ser10)
Clonality: Monoclonal
Clone: 11D8
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR