Anti-Granulysin Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: DH2]

Supplier: Biolegend
348003 348004
348003-BLEA 145.2 USD
348003-BL 348004-BL
Anti-Granulysin Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: DH2]
Antibodies
PE anti-human Granulysin [DH2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application. Read more at www.biolegend.com/testsize regarding the test size change.

Type: Primary
Antigen: Granulysin
Clonality: Monoclonal
Clone: DH2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR