Anti-CD45RB Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: C363-16A]

Supplier: Biolegend
103315 103316
103315-BLEA 145.3 USD
103315-BL 103316-BL
Anti-CD45RB Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: C363-16A]
Antibodies
Pacific Blue™ anti-mouse CD45RB [C363-16A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. The suggested use of this reagent is 1.0 μg per 106 cells in 100 μl volume or 100 μl of whole blood. It is highly recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD45RB
Clonality: Monoclonal
Clone: C363-16A
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR