Anti-IL-2 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: JES6-5H4]

Supplier: Biolegend
503805 503806
503805-BLEA 87.8 USD
503805-BL 503806-BL
Anti-IL-2 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: JES6-5H4]
Antibodies
FITC anti-mouse IL-2 [JES6-5H4]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: IL-2
Clonality: Monoclonal
Clone: JES6-5H4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR