Anti-CD3 Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: UCHT1]

Supplier: Biolegend
300435 300436
300435-BLEA 246.2 USD
300435-BL 10757-108
Anti-CD3 Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: UCHT1]
Antibodies
Brilliant Violet 570™ anti-human CD3 [UCHT1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: UCHT1
Conjugation: Brilliant Violet® 570
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Chimpanzee
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR