Anti-CD19 Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: HIB19]

Supplier: Biolegend
302235 302236
302235-BLEA 229.8 USD
302235-BL 10757-688
Anti-CD19 Mouse Monoclonal Antibody (Brilliant Violet® 570) [clone: HIB19]
Antibodies
Brilliant Violet 570™ anti-human CD19 [HIB19]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: Brilliant Violet® 570
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Chimpanzee;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR