Anti-CD4 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: GK1.5]

Supplier: Biolegend
100437 100438 100443
100437-BLEA 239.2 USD
100437-BL 100438-BL 10756-660
Anti-CD4 Rat Monoclonal Antibody (Brilliant Violet® 421) [clone: GK1.5]
Antibodies
Brilliant Violet 421™ anti-mouse CD4 [GK1.5]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IF; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining using the μg size, the suggested use of this reagent is 0.125 μg per million cells in 100 μl volume. For flow cytometric staining using the μl size, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: GK1.5
Conjugation: Brilliant Violet® 421
Epitope:
Host: Rat
Isotype: Rat IgG2b, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR