Anti-CCL2 Mouse Monoclonal Antibody (Biotin) [clone: 5D3-F7]

Supplier: Biolegend
502608 502609
502608-BLEA 132.9 USD
502608-BL 502609-BL
Anti-CCL2 Mouse Monoclonal Antibody (Biotin) [clone: 5D3-F7]
Antibodies
Biotin anti-human MCP-1 [5D3-F7]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus; Apps: ELISA Detection, ELISPOT Detection, ICFC; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA detection antibody, a concentration range of 0.5-2 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of MCP-1 recombinant protein ranging from 2000 to 15 pg/ml are recommended for each ELISA plate. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CCL2
Clonality: Monoclonal
Clone: 5D3-F7
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR