Anti-IL-4 Rat Monoclonal Antibody (Biotin) [clone: MP4-25D2]

Supplier: Biolegend
500803 500804
500803-BLEA 113.7 USD
500803-BL 500804-BL
Anti-IL-4 Rat Monoclonal Antibody (Biotin) [clone: MP4-25D2]
Antibodies
Biotin anti-human IL-4 [MP4-25D2]; Isotype: Rat IgG1, κ; Reactivity: Human, Cross-Reactivity: Swine (Pig, Porcine), Rhesus; Apps: ELISA Detection, ELISPOT Detection, ICFC; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For use as an ELISA detection antibody, a concentration range of 0.25-1.0 μg/ml is recommended. To obtain a linear standard curve, serial dilutions of human IL-4 protein ranging from 500 to 2 pg/ml are recommended for each ELISA plate.

Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone: MP4-25D2
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Human;Cross-Reactivity: Swine (Pig;Porcine);Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR