Anti-CD90.1 Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: OX-7]

Supplier: Biolegend
202519 202520
202519-BLEA 140.28 USD
202519-BL 202520-BL
Anti-CD90.1 Mouse Monoclonal Antibody (APC (Allophycocyanin)/Cy7®) [clone: OX-7]
Antibodies
APC/Cy7 anti-rat CD90/mouse CD90.1 (Thy-1.1) [OX-7]; Isotype: Mouse IgG1, κ; Reactivity: Rat, Mouse (AKR/J and PL mouse strains). Cross-Reactivity: Rabbit (Lapine), Guinea Pig; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.06 μg per 106 cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD90.1
Clonality: Monoclonal
Clone: OX-7
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Rat;Mouse (AKR/J and PL mouse strains). Cross-Reactivity: Rabbit (Lapine);Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR