Anti-IL-2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MQ1-17H12]
Supplier: Biolegend
500309 500310 500311
500309-BLEA 127.3 USD
500309-BL 500310-BL 500311-BL
Anti-IL-2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MQ1-17H12]
Antibodies
APC anti-human IL-2 [MQ1-17H12]; Isotype: Rat IgG2a, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, Sooty Mangabey; Apps: ICFC; Size: 50 μg
Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. Test size products are transitioning from 20 μl to 5 μl per test. Please check your vial or your CoA to find the suggested use of this reagent per million cells in 100 μl staining volume or per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance forMore Product Information