Anti-IgG1, k Isotype Ctrl (ICFC) Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: MOPC-21]

Supplier: Biolegend
400133 400134
400133-BLEA 93.4 USD
400133-BL 400134-BL
Anti-IgG1, k Isotype Ctrl (ICFC) Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: MOPC-21]
Antibodies
Alexa Fluor® 488 Mouse IgG1, κ Isotype Ctrl (ICFC) [MOPC-21]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: ICFC, IF; Size: 100 tests

Each lot of this mouse IgG1, κ isotype control antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.

Type: Control
Antigen: IgG1, k Isotype Ctrl (ICFC)
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR