Anti-CD146 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: SHM-57]

Supplier: Biolegend
342007 342008
342007-BLEA 145.3 USD
342007-BL 342008-BL
Anti-CD146 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: SHM-57]
Antibodies
Alexa Fluor® 488 anti-human CD146 (MUC18, Mel-CAM) [SHM-57]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD146
Clonality: Monoclonal
Clone: SHM-57
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;African Green;Baboon;Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR