Anti-IgG Goat Polyclonal Antibody (CF™ 640R)

Supplier: Biotium

20085 20085-1
71003-944EA 217.08 USD
71003-944 71003-946
Anti-IgG Goat Polyclonal Antibody (CF™ 640R)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 640R
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR