Anti-IgG Goat Polyclonal Antibody (CF™ 770)

Supplier: Biotium

20077 20077-1
89138-532EA 324.03 USD
89138-532 89138-534
Anti-IgG Goat Polyclonal Antibody (CF™ 770)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 770
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR