Anti-IgG Rabbit Polyclonal Antibody (CF™ 680)

Supplier: Biotium

20243 20243-1
71004-118EA 252.23 USD
71004-118 71004-120
Anti-IgG Rabbit Polyclonal Antibody (CF™ 680)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 680
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Guinea Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR