Anti-IgG Donkey Polyclonal Antibody (CF™ 680)

Supplier: Biotium

20062 20062-1
89138-512EA 252.23 USD
89138-512 89138-514
Anti-IgG Donkey Polyclonal Antibody (CF™ 680)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 680
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR