Anti-IgG Goat Polyclonal Antibody (CF™ 660R)

Supplier: Biotium

20054-1 20054
71003-994EA 86.82 USD
71003-994 89410-838
Anti-IgG Goat Polyclonal Antibody (CF™ 660R)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 660R
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR