Anti-IgY Goat Polyclonal Antibody (CF™ 660C)

Supplier: Biotium

20371-500UL 20371-50UL
89427-262EA 260.49 USD
89427-262 89427-264
Anti-IgY Goat Polyclonal Antibody (CF™ 660C)
Antibodies
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: CF™ 660C
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR