Anti-IgG Goat Polyclonal Antibody (CF™ 660C)

Supplier: Biotium

20052 20052-1
89138-498EA 286.57 USD
89138-498 89138-500
Anti-IgG Goat Polyclonal Antibody (CF™ 660C)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 660C
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR