Anti-IgG2a Goat Polyclonal Antibody (CF™ 647)

Supplier: Biotium

20262 20262-1
71004-248EA 279.11 USD
71004-248 71004-250
Anti-IgG2a Goat Polyclonal Antibody (CF™ 647)
Antibodies
Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: CF™ 647
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR