Anti-IgG Rabbit Polyclonal Antibody (CF™ 647)

Supplier: Biotium

20285 20285-1
89138-824EA 260.49 USD
89138-824 89138-826
Anti-IgG Rabbit Polyclonal Antibody (CF™ 647)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 647
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR