Anti-IgG Goat Polyclonal Antibody (CF™ 647)

Supplier: Biotium

20282 20282-1
89138-812EA 322.57 USD
89138-812 89138-814
Anti-IgG Goat Polyclonal Antibody (CF™ 647)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 647
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR