Anti-IgG Donkey Polyclonal Antibody (CF™ 647)

Supplier: Biotium

20047 20047-1
89138-484EA 330.6 USD
89138-484 89138-486
Anti-IgG Donkey Polyclonal Antibody (CF™ 647)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 647
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR