Anti-IgY Donkey Polyclonal Antibody (CF™ 633)

Supplier: Biotium

20168 20168-1
97065-520EA 322.57 USD
97065-520 97065-522
Anti-IgY Donkey Polyclonal Antibody (CF™ 633)
Antibodies
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: CF™ 633
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR