Anti-IgG Goat Polyclonal Antibody (CF™ 633)

Supplier: Biotium

20130 20130-1
89138-664EA 172.63 USD
89138-664 10018-418
Anti-IgG Goat Polyclonal Antibody (CF™ 633)
Antibodies
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-mouse IgG (H+L) with CF™633 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Far-red fluorescent CF™633 (Ex/Em 630/650 nm) produces brighter antibody conjugates than Alexa Fluor® 647, and has unmatched photostability.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific. Cy®5 is a registered trademark of GE Healthcare.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 633
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR