Anti-IgG1 Goat Polyclonal Antibody (CF™ 594)

Supplier: Biotium

20249 20249-1
71003-858EA 279.11 USD
71003-858 71003-860
Anti-IgG1 Goat Polyclonal Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgG1
Clonality: Polyclonal
Clone:
Conjugation: CF™ 594
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR