Anti-IgY Rabbit Polyclonal Antibody (CF™ 594)

Supplier: Biotium

20164 20164-1
71003-880EA 217.08 USD
71003-880 71003-882
Anti-IgY Rabbit Polyclonal Antibody (CF™ 594)
Antibodies
Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: CF™ 594
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR