Anti-IgG Goat Polyclonal Antibody (CF™ 594)

Supplier: Biotium

20153 20153-1
89138-730EA 172.63 USD
89138-730 10018-258
Anti-IgG Goat Polyclonal Antibody (CF™ 594)
Antibodies
This conjugate is prepared by labeling F(ab’)2 fragment of goat anti-rabbit IgG (H+L) with CF™594 dye. CF™ dyes offer advantages in brightness, photostability, and specificity compared to other fluorescent dyes. Deep-red fluorescent CF™594 (Ex/Em 593/614 nm) is spectrally similar to Alexa Fluor® 594 and Texas Red® dye but yields much brighter protein conjugate, and has excellent photostability.

Alexa Fluor® and Texas Red® are registered trademarks of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 594
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR