Anti-IgG Bovine Polyclonal Antibody (CF™ 568)

Supplier: Biotium

20294 20294-1
71003-928EA 260.49 USD
71003-928 71003-930
Anti-IgG Bovine Polyclonal Antibody (CF™ 568)
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF™ 568
Epitope: Heavy and Light chain
Host: Bovine
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR